Learn More
Medchemexpress LLC (Z)-orantinib | 210644-62-5 | MFCD09743433 | 99.4% | 310.35 g/mol | C18H18N2O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY10517A5MG
Encompass_Preferred
(Z)-Orantinib is the Z-isomer small-molecule inhibitor of VEGFR (Flk-1/KDR), PDGFRβ, and FGFR1 used in preclinical research to investigate angiogenesis and tumor growth inhibition. Supplied as an orange-red solid with high reported purity, it is intended for laboratory research use only. The compound is available in multiple solid pack sizes and as a DMSO solution, with recommended storage conditions for powder and solvent formats.
  • ATP-competitive inhibitor of VEGFR, PDGFRβ, and FGFR1.
  • Reported IC50s: Flk-1/KDR 2.1 μM; PDGFRβ 0.008 μM; FGFR1 1.2 μM.
  • High purity suitable for in vitro biochemical and cellular assays.
  • Available as solid powder and as 10 mM solution in DMSO.
  • Storage conditions provided for powder and solvent preparations.
Catalog No. 50-002-44096