Learn More
Medchemexpress LLC Z-Phe-Ala-diazomethylketone (PADK) | 71732-53-1 | 98.1% | 394.42 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP42955MG
Encompass_Preferred
Z-Phe-Ala-diazomethylketone, also known as PADK, directly binds to Aβ42 monomers and small oligomers. It effectively inhibits the formation of Aβ42 dodecamers and prevents Aβ42 fibril formation in solution. This compound demonstrates significant potential for research into neurodegenerative disorders.
  • Binds directly to Aβ42 monomers and small oligomers
  • Inhibits the formation of Aβ42 dodecamers
  • Inhibits Aβ42 fibril formation in solution
  • Has potential for neurodegenerative disorders research
Catalog No. 50-003-98776