Learn More
Carbosynth LLC. Z-Phe-Arg-MCA (5 mg vial), 1ea, MW: 612.679999999999, Molecular formula: C33H36N6O6, CAS# 65147-22-0
SDP
Supplier:  Carbosynth LLC. MFR3095V5MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: Benzyloxycarbonyl-L-Phenylalanyl-L-Arginine 4-Methyl-Coumaryl-7-Amide, Z-Phe-Arg-AMC, Z-FR-AMC, Z-FR-MCA, Purity: , Grade AA, Application: Substrate for Plasma Kallikrein, Cathespin B/L, and Arg-Gingipain, Salt Form:, Hydrochloride Form, Storage Temperature: -20C, References: T. Morita, H. Kato, S. Iwanaga, K. Takada, T. Kimura, and S. Sakakibara, J. Biochem., 82, 1495 (1977)., A.J. Barret, J. Biochem, 187, 909 (1980).
Catalog No. 50-168-5973