Learn More
Medchemexpress LLC Z-Phe-Arg-pNA | 117761-01-0 | 99.87% | 575.62 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP451510MG
Encompass_Preferred
Z-Phe-Arg-pNA is identified as a substrate for Cathepsin L. It is for research use only and not sold to patients.
  • Substrate of Cathepsin L.
  • For research use only.
Catalog No. 50-003-97683