Learn More
Medchemexpress LLC Z-Phe-Arg-pNA | 117761-01-0 | 99.87% | 575.62 | 25 MG

Supplier: Medchemexpress LLC 25MGHYP4515
Z-Phe-Arg-pNA is a substrate of Cathepsin L. This product is intended for laboratory research and is shipped at room temperature.
- Sealed storage away from moisture
- Powder storage at -80°C for 2 years or -20°C for 1 year
- In solvent storage at -80°C for 6 months or -20°C for 1 month
- H2O solubility: 25 mg/mL (43.43 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.