Learn More
Medchemexpress LLC Z-Phe-Arg-pNA | 117761-01-0 | 99.9% | 575.62 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP45155MG
Encompass_Preferred
Z-Phe-Arg-pNA is a substrate of Cathepsin L.
  • Substrate of Cathepsin L
Catalog No. 50-003-97962