Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-Phe-Leu-OH | 4313-73-9 | MFCD00038301 | 412.48 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0109555G

Encompass_Preferred

Z-Phe-Leu-OH is a chemical compound (MedChemExpress catalog number HY-W010955) intended for laboratory use and the manufacture of substances. It has a molecular formula of C23H28N2O5 and a molecular weight of 412.48. This substance is classified as harmful if swallowed, causes skin and serious eye irritation, and may cause respiratory irritation. Proper handling and storage are crucial, recommending it be kept tightly sealed in a cool, well-ventilated area, away from direct sunlight and ignition sources.

  • Used as a laboratory chemical
  • Suitable for the manufacture of substances
  • Molecular formula: C23H28N2O5
  • Molecular weight: 412.48
  • Stable under recommended storage conditions
  • Store in a cool, well-ventilated area
  • Avoid direct sunlight and sources of ignition

Catalog No. 50-004-53856


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.