Learn More
Medchemexpress LLC Z-Phe-Phe-FMK | 105608-85-3 | 98.3% | 462.51 g·mol⁻1 | C27H27FN2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1418675MG
Encompass_Preferred
Z-FF-FMK (Z-Phe-Phe-FMK) is a peptide-based, irreversible inhibitor of cathepsin L supplied as a dry powder for research use. It is used in biochemical and cellular studies to probe protease activity, apoptotic pathways, and amyloid-related proteolysis, and is typically dissolved in DMSO for experimental assays.
  • Peptide-based irreversible inhibitor of cathepsin L.
  • High purity suitable for research applications.
  • Supplied as a dry powder for long-term storage.
  • Soluble in DMSO; ultrasonic assistance may be required.
  • Stable when stored frozen according to recommended conditions.
  • Suitable for biochemical and cellular assays investigating protease-dependent processes.
Catalog No. 50-002-20990