Learn More
Medchemexpress LLC Z-Phe-Tyr(tBu)-diazomethylketone | 114014-15-2 | MFCD00672349 | 98.0% | 542.63 g/mol | C31H34N4O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1382085MG
Encompass_Preferred
Z-Phe-Tyr(tBu)-diazomethylketone is a peptide-based diazomethylketone inhibitor of cathepsin L used in biochemical and virology research. It inhibits cathepsin L-dependent proteolysis and has been applied to studies of viral disassembly and detection.
  • Potent cathepsin L inhibitor.
  • Used in studies of viral entry and reovirus disassembly.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Multiple milligram-scale sizes, including 5 mg solid.
  • High purity (~98.0%) suitable for research use.
  • Well-characterized: CAS 114014-15-2; MW 542.63 g/mol.
Catalog No. 50-002-22412