Learn More
Medchemexpress LLC Z-pro-pro-cho | 108708-25-4 | 98.0% | 330.38 g·mol⁻¹ | C18H22N2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1344545MG
Encompass_Preferred
Z-Pro-Pro-CHO is a peptide aldehyde inhibitor of prolyl oligopeptidase used as a research reagent for biochemical and enzymatic studies. It selectively inhibits human and Schistosoma mansoni prolyl oligopeptidase and is supplied with supporting documentation for laboratory use.
  • Selective prolyl oligopeptidase inhibition (IC50 0.16 μM human; 0.01 μM Schistosoma mansoni).
  • High purity: 98.0%.
  • Molecular formula C18H22N2O4; molecular weight 330.38 g·mol⁻¹.
  • Supplied in milligram pack sizes suitable for assay development.
  • Certificate of analysis and safety data sheet available.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-22443