Learn More
Medchemexpress LLC Z-pro-pro-CHO | 108708-25-4 | 98.0% | C18H22N2O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13445410MG
Encompass_Preferred
Z-Pro-Pro-CHO is a research-grade prolyl oligopeptidase inhibitor supplied as a high-purity small-molecule solid. It inhibits human prolyl oligopeptidase (IC50 0.16 μM) and Schistosoma mansoni prolyl oligopeptidase (IC50 0.01 μM), and is intended for in vitro biochemical and enzymatic studies.
  • High purity, suitable for research (98.0%).
  • Potent inhibitor of prolyl oligopeptidase (IC50 0.16 μM human; 0.01 μM S. mansoni).
  • Characterized small-molecule with formula C18H22N2O4 and molecular weight 330.38.
  • Available in milligram-scale quantities for screening and assays.
  • Intended for in vitro research use only.
Catalog No. 50-002-61780