Learn More
Medchemexpress LLC (Z)-Pugnac | 132489-69-1 | 99.1% | 353.33 g/mol | C15H19N3O7 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10824110MM 1ML
Encompass_Preferred
(Z)-PUGNAc is the Z stereoisomer of PUGNAc and a potent inhibitor of O-GlcNAcase (OGA) used to increase protein O-GlcNAcylation in biochemical and cellular research. It is available as a solid or as a ready-to-use 10 mM solution in DMSO.
  • Potent O-GlcNAcase inhibitor.
  • Elevates protein O-GlcNAcylation in cells.
  • Available as solid or 10 mM solution in DMSO.
  • High reported purity (99.13%).
  • Molecular weight 353.33 g/mol.
Catalog No. 50-002-44068