Learn More
TARGETMOL CHEMICALS INC Z-PUGNAc 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T387225MG
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (Z)-PUGNAc, a potent O-GlcNAcase inhibitor more active than (E)-PUGNAc, removes O-GlcNAc from proteins (peptide O-GlcNAc-beta-N-acetylaminoglucosidase) and can be used to increase levels of O-GlcNAc on nuclear and cytoplasmic proteins in vivo. Purity 97.58%
Catalog No. 50-287-5944