Learn More
Medchemexpress LLC Z-Pyr-OH (Cbz-L-pyroglutamic acid) | 32159-21-0 | 99.9% | 263.25 | 50 G

Supplier: Medchemexpress LLC HYW01175050G
Z-Pyr-OH (Cbz-L-pyroglutamic acid) is a biochemical assay reagent with a purity of 99.92% and a molecular weight of 263.25. It appears as a white to off-white solid.
- Used as a biochemical assay reagent
- Available as a white to off-white solid
- Purity of 99.92%
- Molecular weight of 263.25
- Related to peptides
- Used in peptide synthesis
- An amino acid derivative
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.