Learn More
Medchemexpress LLC (Z)-semaxanib | 194413-58-6 | MFCD09763655 | 238.29 g/mol | C15H14N2O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY1033225MG
Encompass_Preferred
(Z)-Semaxanib is a small-molecule tyrosine kinase inhibitor supplied for research use as a light yellow to yellow solid. It shows cytotoxic activity against TAMH and HepG2 cell lines (IC50 6.28 μM and 8.17 μM, respectively) and is soluble in DMSO.
Catalog No. 50-002-44931