Learn More
Bachem Z-Ser-OBzl
SDP
Supplier:  Bachem 4004231.0005
Encompass_Preferred
Z-Ser-OBzl, 21209-51-8, C18H19NO5, Mr 329.35, 5g. Educt for the synthesis of racemic Gla derivatives. The versatile intermediate Z-Dha-OBzl can be obtained by treating Z-Ser-OBzl with DCC and CuCl.
Catalog No. 50-313-192