Learn More
TARGETMOL CHEMICALS INC Z-Tenilapine 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T836471MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (Z)-Tenilapine is trans-Tenilapine, cis-Tenilapine has sedative activity. Purity 99.46%
Catalog No. 50-287-9162