Learn More
Medchemexpress LLC Z-thiothixene | 3313-26-6 | MFCD00079574 | 443.63 | C23H29N3O2S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10832425MG
Encompass_Preferred
(Z)-Thiothixene is the Z (cis) isomer of thiothixene provided as an analytical reference standard for research use. It serves as a reference material for method development and qualitative and quantitative analysis by HPLC, GC, and MS, and is typically supplied with batch documentation such as a certificate of analysis, SDS, and spectral data.
  • Identifies as (Z)-thiothixene, CAS 3313-26-6.
  • Molecular formula C23H29N3O2S2; molecular weight 443.63.
  • Intended for analytical applications and method development.
  • Supplied with batch-specific certificate of analysis and supporting spectral data.
  • Suitable for HPLC, GC, and mass spectrometry workflows.
Catalog No. 50-002-43985