Learn More
Medchemexpress LLC Z-Thr(Bzl)-OH | 69863-36-1 | 98.9% | 343.37 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW013198
Encompass_Preferred
Z-Thr(Bzl)-OH is a protected amino acid derivative often used in peptide synthesis and as a building block in various organic chemistry applications. It features a benzyloxycarbonyl (Z) protecting group on the amine and a benzyl ester protecting group on the carboxylic acid, making it suitable for selective deprotection and complex synthesis.
  • Protected amino acid derivative
  • Suitable for peptide synthesis
  • Useful as a chemical building block
  • Features selective protecting groups
Catalog No. 50-003-66687