Learn More
Bachem Z-Tyr-Val-Ala-DL-Asp-fluoromethylketone
SDP
Supplier:  Bachem 4027532.0001
Encompass_Preferred
Z-Tyr-Val-Ala-DL-Asp-fluoromethylketone, 1926163-56-5, C30H37FN4O9, Mr 616.64, 1mg. Z-YVAD-FMK, Z-Tyr-Val-Ala-DL-Asp-FMK. Z-YVAD-FMK, a cell-permeable potent irreversible inhibitor of caspase-1 and -4.
Catalog No. 50-260-557