Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-vad | 162852-62-2 | 95.1% | 437.44 g/mol | C20H27N3O8 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY16438810MG

Encompass_Preferred

Z-VAD is an irreversible, broad-spectrum pan-caspase inhibitor supplied as a 10 mg solid for research use. It inhibits multiple caspases, blocks apoptosis signaling, and may induce autophagy or necrosis in some cell models.

  • Irreversible pan-caspase inhibitor active against caspase-3, -6, -7, -8, and -9.
  • Supplied as a 10 mg solid powder for laboratory research use.
  • Molecular weight 437.44 g/mol; chemical formula C20H27N3O8.
  • CAS number 162852-62-2 for unambiguous identification.
  • Soluble in DMSO (~50 mg/mL); formulation guidance provided for in vivo vehicles.
  • Store sealed and away from moisture; powder: -80°C (2 years) or -20°C (1 year).
  • Not for clinical or diagnostic use; intended for research applications only.

Catalog No. 50-002-89915


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.