Learn More
Selleck Chemical LLC Z-VAD
SDP
Supplier:  Selleck Chemical LLC S810210MM/1ML
Encompass_Preferred
Z-VAD(OH)-FMK (Caspase Inhibitor VI) is an irreversible pan-caspase inhibitor
Catalog No. 50-000-00267