Learn More
Medchemexpress LLC Z-VAD-FMK | 161401-82-7 | 99.4% | 453.46 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY16658B100MG
Encompass_Preferred
Z-VAD-FMK is a pan-caspase inhibitor and an ICE-like protease inhibitor. It inhibits apoptosis by preventing the processing of CPP32 to its active form and limits cryopreservation-induced apoptosis by reducing caspase-3 activity in in vitro produced bovine embryos. It is immunosuppressive in vitro, inhibiting T cell proliferation, and can be used for the study of acute pancreatitis.
  • Pan-caspase and ICE-like protease inhibitor
  • Inhibits apoptosis
  • Reduces caspase-3 activity
  • Immunosuppressive in vitro, inhibiting T cell proliferation
  • Can be used for the study of acute pancreatitis
Catalog No. 50-003-94193