Learn More
Selleck Chemical LLC Z-VAD-FMK 1mg 187389-52-2
SDP
Supplier:  Selleck Chemical LLC S70231MG
Encompass_Preferred
Z-VAD-FMK is a cell-permeable, irreversible pan-caspase inhibitor, blocks all features of apoptosis in THP.1 and Jurkat T-cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4325