Learn More
Selleck Chemical LLC Z-VAD-FMK
SDP
Supplier:  Selleck Chemical LLC S702310MM/1ML
Encompass_Preferred
Z-VAD-FMK (Z-VAD(OMe)-FMK) is a cell-permeable irreversible pan-caspase inhibitor blocks all features of apoptosis in THP 1 and Jurkat T-cells
Catalog No. 50-000-02482