Learn More
Ubpbio Z-VAD-FMK, 5MG
Supplier:  Ubpbio F7111
Encompass_Preferred
Inhibitors – Z-VAD-FMK (UBPBio catalog# F7110, F7111, F7112) – Z-VAD-FMK is a cell-permeable pan caspase inhibitor that irreversibly binds to the catalytic site of caspases and can inhibit induction of apoptosis.
Catalog No. 50-112-6908