Learn More
Enzo Life Sciences Z-VAD(OMe)-FMK (5mg). CAS: 187389-52-2
SDP
Supplier:  Enzo Life Sciences BMLP4160005
Encompass_Preferred
Irreversible and cell permeable broad-spectrum caspase inhibitor. Inhibits Fas-mediated apoptosis in Jurkat T cells and apoptosis in THP.1 cells induced by diverse stimuli (including cycloheximide, thapsigargin, etoposide and staurosporine). For cell free assays use ALX-260-020. Alternative name: Caspase family inhibitor (fluoromethylketone). Purity: ≥95%. Solubility: Soluble in DMSO (>10mg/ml), dimethyl formamide (7mg/ml) or 100% ethanol (1.3mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1347