Learn More
Medchemexpress LLC Z-val-ala-asp | 162852-62-2 | 99.8% | 437.44 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1643885MG
Encompass_Preferred
Z-VAD is an irreversible, broad-spectrum pan-caspase inhibitor that can inhibit a variety of caspases. It blocks apoptosis signaling pathways, induces autophagy and necrosis in tumor cells, and exhibits anti-angiogenic activity. It can also enhance the sensitivity of breast cancer and lung cancer cells to radiotherapy in vitro and in vivo, prolonging the growth delay of tumor xenograft models. Z-VAD is well tolerated and is primarily used in research concerning cancer radiosensitization and cell death pathway regulation.
  • Inhibits a variety of caspases
  • Blocks apoptosis signaling pathways
  • Induces autophagy and necrosis in tumor cells
  • Has anti-angiogenic activity
  • Enhances sensitivity of breast and lung cancer cells to radiotherapy
  • Prolongs growth delay of tumor xenograft models
  • Well tolerated
  • Mainly used in research related to cancer radiosensitization and cell death pathway regulation
Catalog No. 50-003-92422