Learn More
Medchemexpress LLC Z-Val-Gly-OH | 2790-84-3 | 308.33 g/mol | C15H20N2O5 | 1 G

Supplier: Medchemexpress LLC HYW0488301G
Z-Val-Gly-OH is a N-benzyloxycarbonyl (Z)-protected dipeptide used as a research reagent and intermediate in peptide synthesis and analytical studies. Supplied for laboratory research use only; not for human or clinical use.
- N-benzyloxycarbonyl (Z)-protected dipeptide
- Cas number 2790-84-3
- Molecular formula C15H20N2O5
- Molecular weight 308.33 g/mol
- Typical pack size 1 g
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.