Learn More
Carbosynth LLC. Z-Val-Val-Arg-MCA (5 mg vial), 1ea, MW: 663.76, Molecular formula: C34H45N7O7, CAS# 124485-41-2
SDP
Supplier:  Carbosynth LLC. MCA3211V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Benzyloxycarbonyl-L-Valyl-L-Valyl-L-Arginine 4-Methyl-Coumaryl-7-Amide, Z-Val-Val-Arg-AMC, Z-VVR-AMC, Z-VVR-MCA, Purity: , Grade AA, Application: Substrate for Cathepsin S/L, Salt Form:, Hydrochloride Form, Storage Temperature: -20C, References: D Brömme, A. Steinert, S. Friebe, S. Fittkau, B. Wiederanders, and H. Kirschke, Biochem. J., 264, 475 (1989)., H. Kirschke and B. Wiederanders,Proteolytic Enzymes: Serine and Cysteine Peptidases, Methods in Enzymology, Vol. 244, (A.J. Barret, Ed.), Academic Press, New York, 1994, pp. 500-511.
Catalog No. 50-168-5952