Learn More
Medchemexpress LLC Z-VDVAD-FMK | 210344-92-6 | 98.0% | 695.73 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP10081MG
Encompass_Preferred
Z-VDVAD-FMK is a special inhibitor of caspase-2 that produces a reduction in Lovastatin-induced apoptosis.
  • Special inhibitor of caspase-2.
  • Reduces lovastatin-induced apoptosis.
  • Prevents many morphological features of apoptosis in cotreatment.
  • Greatly inhibits Rho-kinase activity in HMEC-1 cells stimulated by thrombin.
  • Significantly reduces lovastatin-induced loss of DNA.
Catalog No. 50-004-24581