Learn More
Apexbio Technology LLC Z-VDVAD-FMK 210344-92-6 5mg
SDP
Supplier:  Apexbio Technology LLC A19225
Encompass_Preferred
Z-VDVAD-FMK (CAS 210344-92-6) is an irreversible cell-permeable inhibitor primarily targeting caspase-2 with additional inhibitory effects on caspases-3 and -7 It functions by binding covalently to the active site of caspase enzymes thereby interrupting downstream apoptotic signaling processes In cellular studies treatment with Z-VDVAD-FMK suppresses caspase-2-dependent cytochrome c release triggered by apoptotic inducers such as etoposide Moreover Z-VDVAD-FMK effectively reduces apoptosis-related responses including cellular detachment DNA fragmentation and cleavage of PARP2 in endothelial cells exposed to oxyhemoglobin This small molecule serves as a pivotal tool for dissecting caspase pathways and investigating apoptosis-mediated cellular outcomes in biomedical research
Catalog No. 50-000-63053