Learn More
Medchemexpress LLC Z-VEID-FMK | 210344-96-0 | 95.0% | 652.71 | 10 MG

Supplier: Medchemexpress LLC HYP100710MG
Z-VEID-FMK (Z-VE(OMe)ID(OMe)-FMK) is a selective and irreversible caspase-6 peptide inhibitor. It alleviates drug-induced augmentation of caspase-6 activity, DNA fragmentation, and cell apoptosis.
- Selective and irreversible caspase-6 peptide inhibitor
- Alleviates drug-induced augmentation of caspase-6 activity
- Reduces DNA fragmentation
- Decreases cell apoptosis
- Pretreatment with 50 μM decreased drug-induced caspase-6 activity by 53% in HepG2 cells
- Pretreatment with 50 μM decreased drug-induced apoptosis by 58% in HepG2 cells
- Alleviates S-(+)-ketamine-caused DNA fragmentation by 44% in HepG2 cells
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.