Learn More
Medchemexpress LLC Z-VEID-FMK | 210344-96-0 | 95.0% | 652.71 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP1007
Encompass_Preferred
Z-VEID-FMK is a selective and irreversible peptide inhibitor of caspase-6. This compound has been shown to alleviate drug-induced increases in caspase-6 activity, DNA fragmentation, and cell apoptosis.
  • Selectively and irreversibly inhibits caspase-6
  • Decreases drug-induced caspase-6 activity, apoptosis, and DNA fragmentation
  • In vitro study showed reduction in caspase-6 activity, apoptosis, and DNA fragmentation in HepG2 cells
  • For research use only
Catalog No. 50-004-26878