Learn More
Medchemexpress LLC Z-VEID-FMK | 210344-96-0 | 95.0% | 652.71 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP10075MG
Encompass_Preferred
Z-VEID-FMK is a selective and irreversible caspase-6 peptide inhibitor. It alleviates drug-induced augmentation of caspase-6 activity, DNA fragmentation, and cell apoptosis.
  • Selective and irreversible caspase-6 peptide inhibitor.
  • Alleviates drug-induced augmentation of caspase-6 activity.
  • Reduces DNA fragmentation.
  • Decreases cell apoptosis.
Catalog No. 50-004-22483