Learn More
Medchemexpress LLC Z-WEHD-FMK | 210345-00-9 | 98.0% | 763.77 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP0111
Encompass_Preferred
Z-WEHD-FMK is a potent, cell-permeable, and irreversible caspase-1/5 inhibitor. It also demonstrates a robust inhibitory effect on cathepsin B activity with an IC50 of 6 μM, and can be utilized to investigate cells for evidence of apoptosis.
  • Potent, cell-permeable, and irreversible caspase-1/5 inhibitor
  • Demonstrates a robust inhibitory effect on cathepsin B activity
  • Utilized to investigate cells for evidence of apoptosis
  • Appears as a light yellow to yellow solid
  • Soluble in DMSO
Catalog No. 50-004-26873