Learn More
Medchemexpress LLC Z-YVAD-AFC | 201608-13-1 | 98.85% | 811.76 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP44291MG
Encompass_Preferred
Z-YVAD-AFC is a fluorescent substrate for Caspase-1, used to detect and measure its activity. It is intended for research use only and not for patient administration.
  • Detects and measures Caspase-1 activity
  • Fluorescent substrate
  • Used as a laboratory chemical
  • Stable under recommended storage conditions
Catalog No. 50-003-98743