Learn More
Medchemexpress LLC Z-YVAD-AFC | 201608-13-1 | 98.9% | 811.76 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP442910MG
Encompass_Preferred
Z-YVAD-AFC is a fluorescent substrate specifically designed for Caspase-1, which allows for the detection and measurement of Caspase-1 activity. It is intended for research use only and not for patient use.
  • Fluorescent substrate for Caspase-1
  • Can be used to detect Caspase-1 activity
  • Can be used to measure Caspase-1 activity
Catalog No. 50-003-97650