Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-YVAD-AFC | 201608-13-1 | 98.9% | 811.76 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYP44295MG

Encompass_Preferred

Z-YVAD-AFC is a fluorescent substrate for Caspase-1, used to detect and measure the activity of Caspase-1. This product is for research use only and not intended for patient use.

  • Fluorescent substrate for Caspase-1
  • Detects and measures Caspase-1 activity
  • Purity of 98.85%
  • Molecular weight of 811.76
  • Appearance: white to off-white solid
  • Sequence: Z-Tyr-Val-Ala-{Asp-AFC}
  • Powder stable for 2 years at -80°C
  • Powder stable for 1 year at -20°C
  • In solvent, stable for 6 months at -80°C
  • In solvent, stable for 1 month at -20°C

Catalog No. 50-003-96650


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.