Learn More
Selleck Chemical LLC Z-YVAD-FMK
SDP
Supplier:  Selleck Chemical LLC S8507100MG
Encompass_Preferred
Z-YVAD-FMK is a cell-permeable and irreversible caspase-1 specific inhibitor with anti-inflammatory and anti-tumor activities
Catalog No. 50-000-01294