Learn More
Enzo Life Sciences Z-YVAD(OMe)-FMK (1mg)
SDP
Supplier:  Enzo Life Sciences ALX260074M001
Encompass_Preferred
Irreversible, cell permeable inhibitor of caspase-1. Inhibits also caspase-4. Alternative name: Caspase 1 inhibitor (fluoromethylketone). Formula: C31H39FN4O9. MW: 630.7. Purity: ≥98% (TLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8288