Learn More
Montana Molecular Human Mu Opioid Receptor (MOR, OPRM, OPRM1) Big Sky BacMam, 15mL
SDP
Supplier:  Montana Molecular Z0729N015
Encompass_Preferred
Human Mu Opioid receptor packaged in BacMam for mammalian cell expression. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. BacMam expression is easily titrateable to suit your experimental needs.
Catalog No. 50-281-5237