Learn More
Montana Molecular Human Serotonin 5-HT2C Receptor (HTR2C, 5HT2CR) BacMam, 30mL
SDP
Supplier:  Montana Molecular Z1800N030
Encompass_Preferred
Use this assay to express the human 5-hydroxytryptamine receptor 2C in your cells of interest. BacMam packaging provides efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells. Importantly, BacMam expression is easily titrateable to suit your experimental needs. This tool can be co-expressed with Montana Molecular's assays for DAG, Calcium, or arrestin to investigate signaling dynamics or receptor activation at the 5-HT2C receptor.
Catalog No. 50-281-5244