Learn More
Medchemexpress LLC Zadavotide guraxetan | 2192281-54-0 | 99.9% | 1498.37 g/mol | C63H92IN11O23 | 100 MG

Supplier: Medchemexpress LLC HY148761100MG
Zadavotide guraxetan is a prostate-specific membrane antigen (PSMA)-targeting small-molecule ligand used as a research precursor for PSMA-directed imaging and radioligand studies, including lutetium-177 conjugation. Intended for research use only.
- PSMA-targeting ligand suitable for radiolabeling and imaging applications.
- Reported high purity supports analytical and radiochemistry workflows.
- Available as a solid in multiple pack sizes, including 100 mg.
- CAS number 2192281-54-0 for unambiguous identification.
- Molecular weight 1498.37 g/mol and chemical formula C63H92IN11O23.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.