Learn More
Apexbio Technology LLC Zafirlukast, 107753-78-6, MFCD00864775, 50 mg
SDP
Supplier:  Apexbio Technology LLC B206850
Encompass_Preferred
MF: C31H33N3O6S, MW: 575.68, Solubility: 23.9mg/mL in DMSO. Zafirlukast is a leukotriene receptor antagonist (LTRA). (IC50=0.6 uM, IC50=7.0 uM for CYP2C9).
Catalog No. 50-101-1361