Learn More
Medchemexpress LLC Zagociguat | 2201048-82-8 | 99.09% | C16H10F4N6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14560750MG
Encompass_Preferred
Zagociguat (CY-6463) is an orally active and BBB-permeable soluble guanylate cyclase (sGC) stimulator. It increases cGMP levels, regulates blood pressure, improves neuronal function, reduces inflammatory responses, and has neuroprotective and cognitive-improving effects. It is used in neurodegenerative disease research.
  • Increases cGMP levels
  • Regulates blood pressure
  • Improves neuronal function
  • Reduces inflammatory responses
  • Exhibits neuroprotective and cognitive-improving effects
Catalog No. 50-003-53320