Learn More
Medchemexpress LLC Zagociguat | 2201048-82-8 | 99.1% | C16H10F4N6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY145607100MG
Encompass_Preferred
Zagociguat (CY-6463) is an orally active and BBB-permeable soluble guanylate cyclase (sGC) stimulator. It increases cGMP levels, regulates blood pressure, improves neuronal function, reduces inflammatory responses, and exerts neuroprotective and cognitive-improving effects. Zagociguat can be used in the research of neurodegenerative diseases. It is for research use only and not sold to patients.
  • Increases cGMP levels
  • Regulates blood pressure
  • Improves neuronal function
  • Reduces inflammatory responses
  • Exerts neuroprotective effects
  • Exerts cognitive-improving effects
  • Useful in neurodegenerative disease research
  • Orally active and BBB-permeable sGC stimulator
Catalog No. 50-003-56384