Learn More
Medchemexpress LLC Zalunfiban dihydrochloride | 2815778-41-5 | 99.1% | 1 MG

Supplier: Medchemexpress LLC HY119350B1MG
Zalunfiban dihydrochloride is a research reagent that acts as a selective platelet glycoprotein IIb/IIIa (αIIbβ3) antagonist, used to study platelet aggregation and myocardial infarction mechanisms. The compound is supplied as the dihydrochloride salt with defined purity, molecular properties, and solubility profiles suitable for in vitro and in vivo research.
- Selective αIIbβ3 antagonist with IC50 ≈ 45 nM.
- High purity (≈99.1%) appropriate for biochemical assays.
- Chemical formula C16H20Cl2N8O2S; molecular weight 459.35 g/mol.
- Soluble in DMSO (10 mg/mL); compatible with common in vivo co-solvent systems.
- Storage guidance: solid at 4°C; store solutions at -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.