Learn More
Medchemexpress LLC Zalutumumab, 10mg | CAS No. : 667901-13-5
SDP
Supplier:  Medchemexpress LLC HYP9915510MG
Encompass_Preferred
Zalutumumab is a high affinity, completely human IgG1 monoclonal antibody targeting EGFR. Zalutumumab binds to domain III of the EGF receptor and acts by blocking the binding of EGF and by sterically interfering with the active conformation of the receptor. Zalutumumab binds with IgG and its Fab fragment with EC50s of 7 and 19 nM, respectively. Zalutumumab can be used for the research of cancer.
Catalog No. NC3397913