Learn More
Apexbio Technology LLC Zanamivir, 100mg. Cas: 139110-80-8 MFCD: MFCD00866966. Influenza A/B virus neuraminidases inhibitor
SDP
Supplier:  Apexbio Technology LLC B2136100
Encompass_Preferred
Zanamivir is a selective inhibitor of influenza A and B virus neuraminidases with IC50 values of 0.64-7.9nM. 139110-80-8. MFCD00866966
Catalog No. 50-101-1551